The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308006.1 | NP_001294935.1 | 7 - 8 | 1436 | 69 |
NM_006033.3 | NP_006024.1 | 8 - 9 | 1658 | 69 | |
XM_005258390.1 | XP_005258447.1 | 8 - 9 | 2475 | 69 | |
XM_011526265.2 | XP_011524567.1 | 7 - 8 | 1466 | 69 | |
XM_011526267.1 | XP_011524569.1 | 8 - 9 | 2045 | 69 | |
XM_017026095.1 | XP_016881584.1 | 8 - 9 | 2157 | 69 | |
GenBank mRNA | AF118767.1 | - | 8 - 9 | 1632 | 69 |
AK291799.1 | - | 8 - 9 | 1632 | 69 | |
AK300333.1 | - | 7 - 8 | 1407 | 69 | |
AK315252.1 | - | 8 - 9 | 1660 | 69 | |
BC060825.1 | - | 8 - 9 | 1629 | 69 |