The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006076.4 | NP_006067.3 | 6 - 7 | 1017 | 58 |
XM_017012112.1 | XP_016867601.1 | 6 - 7 | 958 | 58 | |
GenBank mRNA | AF015042.1 | - | 6 - 7 | 941 | 58 |
AK314954.1 | - | 6 - 7 | 1003 | 58 | |
BC017329.2 | - | 6 - 7 | 969 | 58 |