The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145107.1 | NP_001138579.1 | 20 - 21 | 2673 | 68 |
NM_001145108.1 | NP_001138580.1 | 19 - 20 | 2993 | 68 | |
NM_001145109.1 | NP_001138581.1 | 19 - 20 | 2498 | 68 | |
NM_001145110.1 | NP_001138582.1 | 20 - 21 | 2559 | 68 | |
NM_006159.2 | NP_006150.1 | 20 - 21 | 2600 | 68 | |
XM_005268905.3 | XP_005268962.1 | 20 - 21 | 2753 | 68 | |
XM_011538396.1 | XP_011536698.1 | 21 - 22 | 2549 | 68 | |
XM_017019341.1 | XP_016874830.1 | 20 - 21 | 2929 | 68 | |
XM_017019342.1 | XP_016874831.1 | 20 - 21 | 2612 | 68 | |
XM_017019343.1 | XP_016874832.1 | 20 - 21 | 2768 | 68 | |
XM_017019344.1 | XP_016874833.1 | 21 - 22 | 2564 | 68 | |
GenBank mRNA | AK095637.1 | - | 20 - 21 | 2827 | 68 |
AK295125.1 | - | 20 - 21 | 2559 | 68 | |
AK299277.1 | - | 19 - 20 | 2454 | 68 | |
AK299733.1 | - | 20 - 21 | 2673 | 68 | |
AK309825.1 | - | 20 - 21 | 2443 | 68 | |
AK315960.1 | - | 19 - 20 | 2427 | 68 | |
AK316058.1 | - | 20 - 21 | 2920 | 68 | |
BC020544.1 | - | 19 - 20 | 2700 | 68 | |
D83018.1 | - | 20 - 21 | 2494 | 68 | |
D89629.1 | - | 19 - 20 | 2395 | 68 |