The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006623.3 | NP_006614.2 | 1 - 2 | 364 | 66 |
XM_011541226.1 | XP_011539528.1 | 1 - 2 | 423 | 66 | |
XM_011541231.2 | XP_011539533.1 | 1 - 2 | 225 | 66 | |
GenBank mRNA | AF171237.1 | - | 1 - 2 | 210 | 66 |
AK309662.1 | - | 1 - 2 | 224 | 66 | |
AK310132.1 | - | 1 - 2 | 224 | 66 | |
AK315360.1 | - | 1 - 2 | 285 | 66 | |
BC000303.2 | - | 1 - 2 | 204 | 66 | |
BC001349.2 | - | 1 - 2 | 199 | 66 | |
BC011262.1 | - | 1 - 2 | 273 | 66 | |
BC023235.1 | - | 1 - 2 | 177 | 66 | |
CR456795.1 | - | 1 - 2 | 137 | 66 | |
FJ224347.1 | - | - | 829 | 66 |