The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006627.2 | NP_006618.1 | 3 - 4 | 344 | 68 |
GenBank mRNA | AF001176.2 | - | 3 - 4 | 315 | 68 |
AK314790.1 | - | 3 - 4 | 337 | 68 | |
BC004438.1 | - | 3 - 4 | 304 | 68 | |
BC006098.1 | - | 3 - 4 | 310 | 68 | |
CR536569.1 | - | 2 - 3 | 288 | 68 | |
Y18863.1 | - | 3 - 4 | 313 | 68 |