Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAFD1
Entrez Gene ID
10906
Gene Name
TRAF-type zinc finger domain containing 1
Gene Aliases
FLN29
Chromosome Location
Chr.12: 112125545 - 112153604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143906.1 NP_001137378.1 6 - 7 1468 66
NM_006700.2 NP_006691.1 6 - 7 937 66
GenBank mRNA AB007447.1 - 6 - 7 905 66
AK291331.1 - 6 - 7 922 66
AK295474.1 - 6 - 7 678 66
AK312601.1 - 6 - 7 937 66
BC003553.2 - 5 - 6 863 66

View Full Product Details