The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007255.2 | NP_009186.1 | 1 - 2 | 148 | 85 |
GenBank mRNA | AB028600.1 | - | 1 - 2 | 95 | 85 |
AF142675.1 | - | 1 - 2 | 90 | 85 | |
AK023506.1 | - | 1 - 2 | 91 | 85 | |
AY358578.1 | - | 1 - 2 | 131 | 85 | |
BC007317.2 | - | 1 - 2 | 148 | 85 | |
BC062983.1 | - | 1 - 2 | 115 | 85 | |
BC072403.1 | - | 1 - 2 | 137 | 85 |