The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017524.2 | NP_001017524.1 | 4 - 5 | 695 | 113 |
NM_012239.5 | NP_036371.1 | 4 - 5 | 841 | 113 | |
XM_005252835.1 | XP_005252892.1 | 4 - 5 | 841 | 113 | |
XM_011519956.1 | XP_011518258.1 | 4 - 5 | 1633 | 113 | |
XM_011519957.1 | XP_011518259.1 | 4 - 5 | 1462 | 113 | |
XM_017017428.1 | XP_016872917.1 | 4 - 5 | 687 | 113 | |
XM_017017429.1 | XP_016872918.1 | 4 - 5 | 861 | 113 | |
XM_017017430.1 | XP_016872919.1 | 4 - 5 | 855 | 113 | |
XM_017017431.1 | XP_016872920.1 | 4 - 5 | 586 | 113 | |
GenBank mRNA | AF083108.2 | - | 4 - 5 | 909 | 113 |
AK298572.1 | - | 4 - 5 | 590 | 113 | |
AK299438.1 | - | 4 - 5 | 673 | 113 | |
AK301558.1 | - | 3 - 4 | 641 | 113 | |
AK301979.1 | - | 4 - 5 | 829 | 113 | |
AK316175.1 | - | 4 - 5 | 745 | 113 | |
AL535769.3 | - | 4 - 5 | 676 | 113 | |
BC001042.2 | - | 4 - 5 | 824 | 113 | |
BI755839.1 | - | 4 - 5 | 641 | 113 |