The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203504.2 | NP_987100.1 | 7 - 8 | 1027 | 71 |
XM_017008878.1 | XP_016864367.1 | 7 - 8 | 875 | 71 | |
XM_017008879.1 | XP_016864368.1 | 7 - 8 | 901 | 71 | |
GenBank mRNA | AF051311.1 | - | 6 - 7 | 727 | 71 |
AF053535.1 | - | 7 - 8 | 922 | 71 | |
AK291772.1 | - | 7 - 8 | 957 | 71 | |
BC011731.2 | - | 7 - 8 | 945 | 71 |