The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284331.1 | NP_001271260.1 | 5 - 6 | 814 | 63 |
NM_013284.3 | NP_037416.1 | 5 - 6 | 814 | 63 | |
XM_006715692.2 | XP_006715755.1 | 5 - 6 | 785 | 63 | |
XM_006715696.2 | XP_006715759.1 | 3 - 4 | 515 | 63 | |
XM_011515275.1 | XP_011513577.1 | 5 - 6 | 800 | 63 | |
XM_011515279.1 | XP_011513581.1 | 5 - 6 | 800 | 63 | |
GenBank mRNA | AF176097.1 | - | 5 - 6 | 754 | 63 |
AJ131891.2 | - | 5 - 6 | 757 | 63 | |
AK307634.1 | - | 3 - 4 | 487 | 63 | |
BC062590.1 | - | 5 - 6 | 738 | 63 |