The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127207.1 | NP_001120679.1 | 11 - 12 | 1999 | 82 |
NM_014140.3 | NP_054859.2 | 11 - 12 | 2185 | 82 | |
XM_005246631.2 | XP_005246688.1 | 11 - 12 | 2123 | 82 | |
XM_005246632.1 | XP_005246689.1 | 11 - 12 | 2175 | 82 | |
XM_017004228.1 | XP_016859717.1 | 11 - 12 | 1988 | 82 | |
GenBank mRNA | AF082179.1 | - | 10 - 11 | 1955 | 82 |
AF432223.1 | - | 11 - 12 | 1995 | 82 | |
AK000117.1 | - | 7 - 8 | 855 | 82 | |
AK023780.1 | - | 11 - 12 | 2125 | 82 | |
AL122076.1 | - | 9 - 10 | 1419 | 82 | |
BC016482.1 | - | 10 - 11 | 1946 | 82 | |
BC029994.1 | - | 8 - 9 | 1381 | 82 | |
BC043341.1 | - | 11 - 12 | 1972 | 82 |