The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014207.3 | NP_055022.2 | 2 - 3 | 194 | 114 |
GenBank mRNA | AK292698.1 | - | 2 - 3 | 163 | 114 |
BC027901.1 | - | 2 - 3 | 157 | 114 | |
DB143341.1 | - | 2 - 3 | 194 | 114 | |
X04391.1 | - | 2 - 3 | 163 | 114 |