The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014292.4 | NP_055107.3 | 4 - 5 | 389 | 81 |
GenBank mRNA | AK293035.1 | - | 4 - 5 | 271 | 81 |
BC012111.1 | - | 4 - 5 | 268 | 81 | |
BC064900.1 | - | 4 - 5 | 256 | 81 | |
HY128779.1 | - | 4 - 5 | 389 | 81 |