Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL36B
Entrez Gene ID
27177
Gene Name
interleukin 36, beta
Gene Aliases
FIL1, FIL1-(ETA), FIL1H, FILI-(ETA), IL-1F8, IL-1H2, IL1-ETA, IL1F8, IL1H2
Chromosome Location
Chr.2: 113022091 - 113052867 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014438.4 NP_055253.2 1 - 2 55 82
NM_173178.2 NP_775270.1 1 - 2 55 82
GenBank mRNA AF200494.1 - 1 - 2 58 82
AF201833.1 - 1 - 2 58 82

View Full Product Details