The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014601.3 | NP_055416.2 | 3 - 4 | 759 | 111 |
GenBank mRNA | AF181263.1 | - | 2 - 3 | 547 | 111 |
AF454952.1 | - | 2 - 3 | 564 | 111 | |
AK074701.1 | - | 3 - 4 | 681 | 111 | |
AK296907.1 | - | 4 - 5 | 651 | 111 | |
AK298067.1 | - | 2 - 3 | 222 | 111 | |
AK315548.1 | - | 3 - 4 | 683 | 111 | |
BC014445.1 | - | 3 - 4 | 670 | 111 |