The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014653.3 | NP_055468.2 | 1 - 2 | 461 | 73 |
XM_017020250.1 | XP_016875739.1 | 1 - 2 | 461 | 73 | |
GenBank mRNA | AB018332.2 | - | 1 - 2 | 198 | 73 |
BC110330.1 | - | 1 - 2 | 244 | 73 |