The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142685.1 | NP_001136157.1 | 11 - 12 | 1153 | 90 |
NM_014715.3 | NP_055530.2 | 2 - 3 | 485 | 90 | |
XM_005271736.4 | XP_005271793.2 | 12 - 13 | 2183 | 90 | |
XM_011543072.2 | XP_011541374.1 | 12 - 13 | 1385 | 90 | |
XM_011543073.2 | XP_011541375.2 | 12 - 13 | 1121 | 90 | |
XM_011543075.2 | XP_011541377.1 | 3 - 4 | 331 | 90 | |
XM_017018595.1 | XP_016874084.1 | 12 - 13 | 1102 | 90 | |
XM_017018596.1 | XP_016874085.1 | 10 - 11 | 1379 | 90 | |
XM_017018597.1 | XP_016874086.1 | 12 - 13 | 2192 | 90 | |
GenBank mRNA | AB018255.2 | - | 2 - 3 | 406 | 90 |
AB079856.1 | - | 2 - 3 | 485 | 90 | |
AB088416.1 | - | 2 - 3 | 406 | 90 | |
AL833062.2 | - | 10 - 11 | 955 | 90 | |
AY194287.1 | - | 2 - 3 | 409 | 90 | |
BC000277.1 | - | 4 - 5 | 399 | 90 | |
BC051236.1 | - | 6 - 7 | 609 | 90 | |
BC065371.1 | - | 6 - 7 | 609 | 90 | |
BC104898.1 | - | 1 - 2 | 140 | 90 | |
BC113429.1 | - | 1 - 2 | 140 | 90 | |
BQ319051.1 | - | 2 - 3 | 90 | ||
EF127492.1 | - | 11 - 12 | 1153 | 90 |