The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014770.3 | NP_055585.1 | 1 - 2 | 253 | 65 |
XM_005268626.2 | XP_005268683.1 | 1 - 2 | 293 | 65 | |
GenBank mRNA | AF384128.1 | - | 1 - 2 | 249 | 65 |
AF413077.1 | - | 1 - 2 | 163 | 65 | |
AK292672.1 | - | 1 - 2 | 225 | 65 | |
BC028020.1 | - | 1 - 2 | 220 | 65 | |
D79989.1 | - | 1 - 2 | 251 | 65 | |
DB165417.1 | - | 1 - 2 | 253 | 65 |