Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MED24
Entrez Gene ID
9862
Gene Name
mediator complex subunit 24
Gene Aliases
ARC100, CRSP100, CRSP4, DRIP100, MED5, THRAP4, TRAP100
Chromosome Location
Chr.17: 40019097 - 40054636 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079518.1 NP_001072986.1 18 - 19 2282 107
NM_001267797.1 NP_001254726.1 18 - 19 2260 107
NM_014815.3 NP_055630.2 19 - 20 2299 107
NR_052017.1 18 - 19 2225 107
XM_005257874.2 XP_005257931.1 19 - 20 2204 107
XM_006722204.2 XP_006722267.1 19 - 20 2240 107
XM_006722206.2 XP_006722269.1 19 - 20 2148 107
XM_006722207.2 XP_006722270.1 19 - 20 2116 107
XM_011525529.2 XP_011523831.1 20 - 21 2297 107
XM_011525530.2 XP_011523832.1 20 - 21 2137 107
XM_011525531.2 XP_011523833.1 19 - 20 2080 107
XM_011525532.2 XP_011523834.1 20 - 21 2205 107
XM_011525533.2 XP_011523835.1 20 - 21 2173 107
XM_011525534.2 XP_011523836.1 19 - 20 2116 107
XM_011525535.2 XP_011523837.1 19 - 20 2116 107
XM_011525536.2 XP_011523838.1 20 - 21 2137 107
XM_011525538.2 XP_011523840.1 19 - 20 2431 107
XM_017025458.1 XP_016880947.1 20 - 21 2169 107
XM_017025459.1 XP_016880948.1 19 - 20 2112 107
XM_017025460.1 XP_016880949.1 20 - 21 2169 107
XM_017025461.1 XP_016880950.1 20 - 21 2261 107
XM_017025462.1 XP_016880951.1 20 - 21 2169 107
XM_017025463.1 XP_016880952.1 20 - 21 2137 107
XM_017025464.1 XP_016880953.1 19 - 20 2116 107
XM_017025465.1 XP_016880954.1 19 - 20 2240 107
XM_017025466.1 XP_016880955.1 19 - 20 2112 107
XM_017025467.1 XP_016880956.1 19 - 20 2112 107
XM_017025468.1 XP_016880957.1 19 - 20 2080 107
XM_017025469.1 XP_016880958.1 19 - 20 2395 107
XM_017025470.1 XP_016880959.1 19 - 20 2395 107
GenBank mRNA AF055995.1 - 18 - 19 1986 107
AF277379.1 - 18 - 19 1986 107
AK022508.1 - 18 - 19 2051 107
AK291040.1 - 18 - 19 2031 107
AK293309.1 - 17 - 18 1857 107
AK299864.1 - 19 - 20 1916 107
AK300992.1 - 18 - 19 1904 107
AK303502.1 - 7 - 8 754 107
AK303745.1 - 18 - 19 1995 107
BC011375.1 - 18 - 19 2261 107
D50920.1 - 19 - 20 2059 107
EU392542.1 - 4 - 5 344 107

View Full Product Details