The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242307.1 | NP_001229236.1 | 2 - 3 | 230 | 68 |
NM_014889.3 | NP_055704.2 | 2 - 3 | 230 | 68 | |
XM_005252345.3 | XP_005252402.1 | 2 - 3 | 230 | 68 | |
XM_017015470.1 | XP_016870959.1 | 2 - 3 | 298 | 68 | |
XM_017015471.1 | XP_016870960.1 | 2 - 3 | 298 | 68 | |
XM_017015472.1 | XP_016870961.1 | 2 - 3 | 171 | 68 | |
XM_017015473.1 | XP_016870962.1 | 2 - 3 | 171 | 68 | |
GenBank mRNA | AF061243.1 | - | 2 - 3 | 174 | 68 |
AK002061.1 | - | 2 - 3 | 198 | 68 | |
AK225084.1 | - | 2 - 3 | 168 | 68 | |
AK293788.1 | - | 2 - 3 | 168 | 68 | |
AK294865.1 | - | - | 143 | 68 | |
AK299464.1 | - | 2 - 3 | 171 | 68 | |
AK315117.1 | - | 2 - 3 | 168 | 68 | |
BC005025.1 | - | 2 - 3 | 165 | 68 | |
BC095422.1 | - | 2 - 3 | 171 | 68 | |
BC111987.1 | - | 2 - 3 | 186 | 68 | |
BC113369.1 | - | 2 - 3 | 186 | 68 | |
BG720390.1 | - | 2 - 3 | 213 | 68 | |
BI766649.1 | - | 2 - 3 | 153 | 68 | |
BX649017.1 | - | 2 - 3 | 172 | 68 | |
DA331909.1 | - | 2 - 3 | 231 | 68 |