The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079673.1 | NP_001073141.1 | 25 - 26 | 3593 | 109 |
NM_001278438.1 | NP_001265367.1 | 25 - 26 | 3636 | 109 | |
NM_014923.4 | NP_055738.3 | 23 - 24 | 3217 | 109 | |
NR_103528.1 | 25 - 26 | 3625 | 109 | ||
XM_011534997.2 | XP_011533299.1 | 24 - 25 | 3307 | 109 | |
XM_017020440.1 | XP_016875929.1 | 25 - 26 | 3478 | 109 | |
XM_017020441.1 | XP_016875930.1 | 27 - 28 | 4099 | 109 | |
XM_017020442.1 | XP_016875931.1 | 26 - 27 | 3631 | 109 | |
GenBank mRNA | AB023187.2 | - | 23 - 24 | 3147 | 109 |
AJ749706.1 | - | 25 - 26 | 3595 | 109 | |
AJ749707.1 | - | 23 - 24 | 3208 | 109 | |
AK172814.1 | - | 3 - 4 | 524 | 109 | |
AK302415.1 | - | 23 - 24 | 3217 | 109 | |
BC060816.1 | - | 16 - 17 | 2255 | 109 | |
BC070072.1 | - | 16 - 17 | 2255 | 109 | |
BC132812.1 | - | 25 - 26 | 3360 | 109 | |
BC136617.1 | - | 25 - 26 | 3361 | 109 | |
BC144301.1 | - | 25 - 26 | 3361 | 109 | |
BX640739.1 | - | 25 - 26 | 3496 | 109 | |
BX648141.1 | - | 25 - 26 | 3583 | 109 | |
BX648706.1 | - | 27 - 28 | 3441 | 109 |