Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHA6
Entrez Gene ID
22874
Gene Name
pleckstrin homology domain containing A6
Gene Aliases
PEPP-3, PEPP3
Chromosome Location
Chr.1: 204218851 - 204378376 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014935.4 NP_055750.2 15 - 16 2501 85
XM_005244967.4 XP_005245024.2 15 - 16 2427 85
XM_005244968.4 XP_005245025.2 15 - 16 2367 85
XM_006711217.3 XP_006711280.2 18 - 19 2650 85
GenBank mRNA AB023186.1 - 15 - 16 2488 85
BC152475.1 - 15 - 16 2488 85

View Full Product Details