Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP68
Entrez Gene ID
23177
Gene Name
centrosomal protein 68
Gene Aliases
KIAA0582
Chromosome Location
Chr.2: 65056361 - 65087008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319100.1 NP_001306029.1 5 - 6 2312 74
NM_001319101.1 NP_001306030.1 5 - 6 1905 74
NM_015147.2 NP_055962.2 5 - 6 2316 74
GenBank mRNA AB011154.2 - 5 - 6 2316 74
AK000856.1 - 5 - 6 2261 74
AK093846.1 - 4 - 5 1466 74
AK098786.1 - 5 - 6 1277 74
AK299373.1 - 5 - 6 2257 74
BC002982.1 - 5 - 6 1801 74
BC004873.2 - 5 - 6 1801 74

View Full Product Details