Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCL2L13
Entrez Gene ID
23786
Gene Name
BCL2 like 13
Gene Aliases
BCL-RAMBO, Bcl2-L-13, MIL1
Chromosome Location
Chr.22: 17628855 - 17730855 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270726.1 NP_001257655.1 3 - 4 573 77
NM_001270727.1 NP_001257656.1 3 - 4 573 77
NM_015367.3 NP_056182.2 4 - 5 744 77
XM_011546119.1 XP_011544421.1 4 - 5 1107 77
XM_011546120.1 XP_011544422.1 4 - 5 1109 77
XM_011546121.2 XP_011544423.1 5 - 6 1092 77
XM_017028725.1 XP_016884214.1 4 - 5 1107 77
XM_017028726.1 XP_016884215.1 6 - 7 1393 77
XM_017028727.1 XP_016884216.1 5 - 6 686 77
XM_017028733.1 XP_016884222.1 4 - 5 744 77
GenBank mRNA AF146568.1 - 1 - 2 165 77
AF246665.1 - 5 - 6 1130 77
AF325209.1 - 3 - 4 391 77
AK314489.1 - 4 - 5 571 77
BC007658.2 - 4 - 5 503 77
BX407004.2 - 3 - 4 326 77
CR456521.1 - 4 - 5 647 77
DA478149.1 - 3 - 4 403 77

View Full Product Details