Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARM1
Entrez Gene ID
25849
Gene Name
prostate androgen-regulated mucin-like protein 1
Gene Aliases
Cipar1, DKFZP564O0823, PARM-1, WSC4
Chromosome Location
Chr.4: 74933075 - 75050115 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015393.3 NP_056208.2 2 - 3 1022 80
XM_011531833.1 XP_011530135.1 3 - 4 1086 80
GenBank mRNA AK022311.1 - 2 - 3 981 80
AL080121.1 - 2 - 3 938 80
AY358777.1 - 2 - 3 967 80
BC013294.2 - 2 - 3 951 80

View Full Product Details