Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RWDD3
Entrez Gene ID
25950
Gene Name
RWD domain containing 3
Gene Aliases
RSUME
Chromosome Location
Chr.1: 95234155 - 95247225 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128142.1 NP_001121614.1 1 - 2 159 107
NM_001199682.1 NP_001186611.1 1 - 2 159 107
NM_015485.4 NP_056300.2 1 - 2 159 107
NR_103483.1 1 - 2 159 107
GenBank mRNA AK292665.1 - 1 - 2 111 107
AK292699.1 - 1 - 2 159 107
AL050062.1 - 1 - 2 103 107
BC010936.2 - 1 - 2 95 107
CN306675.1 - 1 - 2 94 107
CR533493.1 - 1 - 2 83 107
DC409838.1 - 1 - 2 160 108

View Full Product Details