The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142541.2 | NP_001136013.1 | 6 - 7 | 854 | 61 |
NM_015948.4 | NP_057032.2 | 6 - 7 | 1216 | 61 | |
NR_109913.1 | 6 - 7 | 1065 | 61 | ||
NR_109914.1 | 4 - 5 | 1150 | 61 | ||
XM_005249156.3 | XP_005249213.1 | 5 - 6 | 996 | 61 | |
XM_005249157.2 | XP_005249214.1 | 6 - 7 | 989 | 61 | |
XM_005249158.1 | XP_005249215.1 | 5 - 6 | 884 | 61 | |
XM_006715102.3 | XP_006715165.1 | 6 - 7 | 1026 | 61 | |
XM_006715103.3 | XP_006715166.1 | 5 - 6 | 996 | 61 | |
XM_011514648.1 | XP_011512950.1 | 7 - 8 | 996 | 61 | |
XM_017010910.1 | XP_016866399.1 | 6 - 7 | 1312 | 61 | |
XM_017010911.1 | XP_016866400.1 | 5 - 6 | 1230 | 61 | |
XM_017010912.1 | XP_016866401.1 | 6 - 7 | 944 | 61 | |
XM_017010913.1 | XP_016866402.1 | 6 - 7 | 1312 | 61 | |
XM_017010914.1 | XP_016866403.1 | 5 - 6 | 1080 | 61 | |
XM_017010915.1 | XP_016866404.1 | 5 - 6 | 884 | 61 | |
GenBank mRNA | AB231931.1 | - | 4 - 5 | 691 | 61 |
AF132953.1 | - | 5 - 6 | 712 | 61 | |
AK313522.1 | - | 6 - 7 | 832 | 61 | |
BC006973.1 | - | 6 - 7 | 1042 | 61 | |
BX538271.1 | - | 6 - 7 | 1060 | 61 | |
BX641086.1 | - | 4 - 5 | 6213 | 61 | |
BX647208.1 | - | 4 - 5 | 1084 | 61 | |
BX648022.1 | - | 5 - 6 | 5153 | 61 |