Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPH5
Entrez Gene ID
51611
Gene Name
diphthamide biosynthesis 5
Gene Aliases
AD-018, CGI-30, HSPC143, NPD015
Chromosome Location
Chr.1: 100989623 - 101025806 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077394.1 NP_001070862.1 2 - 3 302 75
NM_001077395.1 NP_001070863.1 2 - 3 284 75
NM_015958.2 NP_057042.2 2 - 3 284 75
XM_005270938.1 XP_005270995.1 2 - 3 284 75
XM_005270941.3 XP_005270998.1 2 - 3 219 75
XM_011541569.1 XP_011539871.1 2 - 3 284 75
XM_017001460.1 XP_016856949.1 2 - 3 304 75
XM_017001462.1 XP_016856951.1 2 - 3 241 75
GenBank mRNA AF132964.1 - 2 - 3 253 75
AF157319.1 - 2 - 3 252 75
AF161492.1 - 2 - 3 293 75
AF248965.1 - 2 - 3 253 75
AK225469.1 - 2 - 3 262 75
AK225707.1 - 2 - 3 262 75
AK225951.1 - 2 - 3 262 75
AK289351.1 - 2 - 3 273 75
AK298181.1 - 2 - 3 262 75
BC053857.1 - 2 - 3 250 75
BI907367.1 - 2 - 3 257 75
CK002046.1 - 2 - 3 305 75
DA133565.1 - 2 - 3 284 75

View Full Product Details