The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015980.4 | NP_057064.1 | 2 - 3 | 394 | 76 |
GenBank mRNA | AF113537.1 | - | 2 - 3 | 263 | 76 |
AF123767.1 | - | 2 - 3 | 263 | 76 | |
AF133422.1 | - | 2 - 3 | 247 | 76 | |
AK297196.1 | - | 2 - 3 | 278 | 76 | |
AK312359.1 | - | 2 - 3 | 281 | 76 | |
BC002619.2 | - | 2 - 3 | 277 | 76 | |
BC018873.2 | - | 2 - 3 | 270 | 76 | |
CR457142.1 | - | 1 - 2 | 132 | 76 | |
DC366641.1 | - | 2 - 3 | 394 | 76 |