The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206651.1 | NP_001193580.1 | 2 - 3 | 544 | 147 |
NM_001206652.1 | NP_001193581.1 | 2 - 3 | 544 | 147 | |
NM_016009.4 | NP_057093.1 | 2 - 3 | 544 | 147 | |
XM_006710672.2 | XP_006710735.1 | 2 - 3 | 544 | 147 | |
XM_011541538.1 | XP_011539840.1 | 2 - 3 | 544 | 147 | |
XM_017001409.1 | XP_016856898.1 | 2 - 3 | 544 | 147 | |
XM_017001410.1 | XP_016856899.1 | 2 - 3 | 544 | 147 | |
GenBank mRNA | AB007960.2 | - | 2 - 3 | 509 | 147 |
AF151819.1 | - | 2 - 3 | 239 | 147 | |
AF257318.1 | - | 2 - 3 | 239 | 147 | |
AF263293.1 | - | 2 - 3 | 219 | 147 | |
AF350371.1 | - | 2 - 3 | 295 | 147 | |
AK001954.1 | - | 2 - 3 | 329 | 147 | |
AK225864.1 | - | 2 - 3 | 364 | 147 | |
BC007455.2 | - | 2 - 3 | 259 | 147 |