The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016141.3 | NP_057225.2 | 5 - 6 | 843 | 127 |
GenBank mRNA | AF078849.1 | - | 5 - 6 | 819 | 127 |
AK001081.1 | - | 2 - 3 | 339 | 127 | |
AK074348.1 | - | 5 - 6 | 827 | 127 | |
AK222573.1 | - | 5 - 6 | 778 | 127 | |
AK222653.1 | - | 5 - 6 | 779 | 127 | |
AK293253.1 | - | 3 - 4 | 479 | 127 | |
BC131620.1 | - | 5 - 6 | 789 | 127 | |
BX649189.1 | - | 5 - 6 | 662 | 127 |