The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001302826.1 | NP_001289755.1 | 7 - 8 | 1148 | 81 |
NM_016229.4 | NP_057313.2 | 7 - 8 | 802 | 81 | |
NR_126508.1 | 7 - 8 | 802 | 81 | ||
XM_005252975.4 | XP_005253032.1 | 7 - 8 | 754 | 81 | |
XM_006718251.2 | XP_006718314.1 | 7 - 8 | 779 | 81 | |
XM_011520184.2 | XP_011518486.1 | 7 - 8 | 969 | 81 | |
XM_011520185.2 | XP_011518487.1 | 9 - 10 | 1068 | 81 | |
XM_017017920.1 | XP_016873409.1 | 9 - 10 | 1062 | 81 | |
GenBank mRNA | AF169802.1 | - | 7 - 8 | 705 | 81 |
AK290102.1 | - | 7 - 8 | 1121 | 81 | |
AL133582.1 | - | 6 - 7 | 538 | 81 | |
AY665398.1 | - | 6 - 7 | 556 | 81 | |
BC001346.1 | - | 7 - 8 | 676 | 81 | |
BF061336.1 | - | 2 - 3 | 81 |