The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130713.2 | NP_001124185.1 | 1 - 2 | 1406 | 60 |
NM_001130714.2 | NP_001124186.1 | 1 - 2 | 1406 | 60 | |
NM_016269.4 | NP_057353.1 | 1 - 2 | 1406 | 60 | |
XM_005263046.3 | XP_005263103.1 | 1 - 2 | 410 | 60 | |
GenBank mRNA | AF198532.1 | - | 1 - 2 | 217 | 60 |
AF288571.1 | - | 1 - 2 | 871 | 60 | |
AF294627.1 | - | 1 - 2 | 217 | 60 | |
AK225772.1 | - | 1 - 2 | 1294 | 60 | |
AK294395.1 | - | 1 - 2 | 248 | 60 | |
BC040559.1 | - | 1 - 2 | 239 | 60 | |
BC050632.2 | - | 1 - 2 | 308 | 60 |