Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRRX2
Entrez Gene ID
51450
Gene Name
paired related homeobox 2
Gene Aliases
PMX2, PRX2
Chromosome Location
Chr.9: 129665641 - 129722674 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016307.3 NP_057391.1 3 - 4 854 80
XM_017014803.1 XP_016870292.1 3 - 4 462 80
GenBank mRNA AF061970.1 - 3 - 4 657 80
BC014645.1 - 3 - 4 854 80
U81600.1 - 2 - 3 360 80

View Full Product Details