Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUSP13
Entrez Gene ID
51207
Gene Name
dual specificity phosphatase 13
Gene Aliases
BEDP, DUSP13A, DUSP13B, MDSP, SKRP4, TMDP
Chromosome Location
Chr.10: 75094432 - 75109221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB027004.1 - 1 - 2 81 100
AB103375.1 - 1 - 2 81 100

View Full Product Details