The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278603.1 | NP_001265532.1 | 10 - 11 | 1394 | 94 |
NM_017426.3 | NP_059122.2 | 11 - 12 | 1538 | 94 | |
NR_103780.1 | 10 - 11 | 1311 | 94 | ||
NR_103781.1 | 9 - 10 | 1203 | 94 | ||
NR_103782.1 | 11 - 12 | 1308 | 94 | ||
XM_006714237.1 | XP_006714300.1 | 12 - 13 | 1515 | 94 | |
XM_011532033.2 | XP_011530335.1 | 10 - 11 | 1373 | 94 | |
XM_011532034.2 | XP_011530336.1 | 10 - 11 | 1781 | 94 | |
XM_017008294.1 | XP_016863783.1 | 11 - 12 | 1925 | 94 | |
XM_017008295.1 | XP_016863784.1 | 10 - 11 | 1698 | 94 | |
GenBank mRNA | AF157322.1 | - | 11 - 12 | 1416 | 94 |
AK001517.1 | - | 11 - 12 | 1420 | 94 | |
AK222752.1 | - | 11 - 12 | 1425 | 94 | |
AK223352.1 | - | 11 - 12 | 1421 | 94 | |
AK299442.1 | - | 10 - 11 | 1193 | 94 | |
AK300036.1 | - | 10 - 11 | 1276 | 94 | |
AK307571.1 | - | 11 - 12 | 1198 | 94 | |
AK315160.1 | - | 11 - 12 | 1418 | 94 | |
AL137650.1 | - | 3 - 4 | 837 | 94 | |
BC012559.1 | - | 11 - 12 | 1435 | 94 | |
BX538002.1 | - | 9 - 10 | 1103 | 94 |