The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032853.3 | NP_116242.2 | 13 - 14 | 2185 | 75 |
NR_024247.1 | 13 - 14 | 2254 | 75 | ||
XM_011528385.2 | XP_011526687.1 | 14 - 15 | 2376 | 75 | |
GenBank mRNA | AK027774.1 | - | 4 - 5 | 654 | 75 |
AK075241.1 | - | 7 - 8 | 815 | 75 | |
AL360266.1 | - | 1 - 2 | 65 | 75 | |
BC008098.1 | - | 5 - 6 | 669 | 75 | |
BC082987.1 | - | 4 - 5 | 636 | 75 | |
BC110874.1 | - | 12 - 13 | 2151 | 75 | |
BC130443.1 | - | 13 - 14 | 2214 | 75 | |
BC144138.1 | - | 13 - 14 | 2258 | 75 | |
BC144147.1 | - | 13 - 14 | 2214 | 75 |