The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017746.3 | NP_060216.2 | 13 - 14 | 2685 | 113 |
XM_011518798.1 | XP_011517100.1 | 11 - 12 | 2272 | 113 | |
XM_017014848.1 | XP_016870337.1 | 13 - 14 | 2174 | 113 | |
GenBank mRNA | AB060968.1 | - | 13 - 14 | 2601 | 113 |
AJ420436.1 | - | 8 - 9 | 1110 | 113 | |
AK000294.1 | - | 13 - 14 | 2601 | 113 | |
AK298916.1 | - | 15 - 16 | 2208 | 113 | |
AY598337.1 | - | 12 - 13 | 2470 | 113 | |
BC030652.2 | - | 13 - 14 | 2521 | 113 | |
BF981005.1 | - | 1 - 2 | 194 | 113 |