The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305544.1 | NP_001292473.1 | 3 - 4 | 1126 | 95 |
NM_001305545.1 | NP_001292474.1 | 2 - 3 | 208 | 95 | |
NM_017763.5 | NP_060233.3 | 3 - 4 | 1329 | 95 | |
XM_011524955.2 | XP_011523257.1 | 3 - 4 | 1228 | 95 | |
XM_011524956.2 | XP_011523258.1 | 2 - 3 | 2005 | 95 | |
XM_017024800.1 | XP_016880289.1 | 4 - 5 | 2555 | 95 | |
GenBank mRNA | AB081837.1 | - | 3 - 4 | 866 | 95 |
AK000322.1 | - | 2 - 3 | 546 | 95 | |
AK291027.1 | - | 3 - 4 | 1099 | 95 | |
AK299024.1 | - | 2 - 3 | 208 | 95 | |
BC109028.2 | - | 2 - 3 | 575 | 95 | |
CR627423.1 | - | 3 - 4 | 1126 | 95 | |
CR749257.1 | - | 2 - 3 | 2334 | 95 |