The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017805.2 | NP_060275.2 | 10 - 11 | 2739 | 121 |
XM_011527053.2 | XP_011525355.2 | 10 - 11 | 2859 | 121 | |
XM_011527054.2 | XP_011525356.2 | 10 - 11 | 2859 | 121 | |
XM_011527055.2 | XP_011525357.2 | 7 - 8 | 2565 | 121 | |
XM_017026914.1 | XP_016882403.1 | 10 - 11 | 2859 | 121 | |
XM_017026915.1 | XP_016882404.1 | 7 - 8 | 2565 | 121 | |
GenBank mRNA | AK000408.1 | - | 6 - 7 | 854 | 121 |
AK123179.1 | - | 2 - 3 | 1036 | 121 | |
AY378097.1 | - | 10 - 11 | 2604 | 121 | |
BC021860.1 | - | 6 - 7 | 869 | 121 | |
BC028614.1 | - | 10 - 11 | 2611 | 121 | |
BX389914.2 | - | 2 - 3 | 238 | 121 |