Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C19orf24
Entrez Gene ID
55009
Gene Name
chromosome 19 open reading frame 24
Gene Aliases
-
Chromosome Location
Chr.19: 1275521 - 1279244 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017914.3 NP_060384.3 2 - 3 421 115
GenBank mRNA AK000647.1 - 2 - 3 194 115
AK096105.1 - 1 - 2 1201 115
AK225502.1 - 2 - 3 194 115
BC000890.1 - 2 - 3 390 115
BG333480.1 - 2 - 3 401 115
BM675280.1 - 1 - 2 115

View Full Product Details