Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIH1D1
Entrez Gene ID
55011
Gene Name
PIH1 domain containing 1
Gene Aliases
NOP17
Chromosome Location
Chr.19: 49446293 - 49452332 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017916.2 NP_060386.1 3 - 4 621 82
GenBank mRNA AK000650.1 - 3 - 4 573 82
AK304476.1 - 3 - 4 672 82
BC001108.2 - 3 - 4 551 82

View Full Product Details