Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFT57
Entrez Gene ID
55081
Gene Name
intraflagellar transport 57
Gene Aliases
ESRRBL1, HIPPI, MHS4R2
Chromosome Location
Chr.3: 108160812 - 108222570 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018010.3 NP_060480.1 10 - 11 1361 105
GenBank mRNA AB014762.1 - 10 - 11 1113 105
AF139576.1 - 10 - 11 1191 105
AF245220.1 - 10 - 11 1255 105
AK001009.1 - 10 - 11 1200 105
AK298854.1 - 10 - 11 1259 105
BC009690.2 - 6 - 7 528 105
BC011899.2 - 10 - 11 1210 105
CR457251.1 - 10 - 11 1113 105

View Full Product Details