Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHG6
Entrez Gene ID
55200
Gene Name
pleckstrin homology and RhoGEF domain containing G6
Gene Aliases
MyoGEF
Chromosome Location
Chr.12: 6310436 - 6328506 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144856.1 NP_001138328.1 2 - 3 365 150
NM_018173.3 NP_060643.2 2 - 3 283 150
XM_005253704.4 XP_005253761.1 2 - 3 380 150
XM_006718985.3 XP_006719048.1 2 - 3 380 150
XM_017019555.1 XP_016875044.1 2 - 3 380 150
GenBank mRNA AK001527.1 - 2 - 3 284 150
AK023741.1 - 2 - 3 365 150
BC104751.1 - 2 - 3 219 150
BC105961.1 - 2 - 3 219 150
BC109095.2 - 2 - 3 219 150

View Full Product Details