The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018180.2 | NP_060650.2 | 1 - 2 | 778 | 89 |
XM_017016404.1 | XP_016871893.1 | 3 - 4 | 1110 | 89 | |
XM_017016405.1 | XP_016871894.1 | 2 - 3 | 800 | 89 | |
GenBank mRNA | AF427340.1 | - | 1 - 2 | 779 | 89 |
AK001556.1 | - | 1 - 2 | 572 | 89 | |
AK001751.1 | - | 1 - 2 | 751 | 89 | |
AK315430.1 | - | 1 - 2 | 393 | 89 | |
AY064250.1 | - | 1 - 2 | 746 | 89 | |
BC002473.2 | - | 1 - 2 | 314 | 89 | |
BC068471.1 | - | 2 - 3 | 775 | 89 |