Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM19
Entrez Gene ID
55266
Gene Name
transmembrane protein 19
Gene Aliases
-
Chromosome Location
Chr.12: 71686098 - 71704060 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018279.3 NP_060749.2 2 - 3 830 99
GenBank mRNA AK001798.1 - 2 - 3 432 99
AK223148.1 - 2 - 3 428 99
AK315587.1 - 2 - 3 389 99
BC008596.1 - 2 - 3 830 99

View Full Product Details