The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018372.3 | NP_060842.3 | 2 - 3 | 1852 | 113 |
XM_005271029.3 | XP_005271086.1 | 2 - 3 | 2111 | 113 | |
XM_011541768.1 | XP_011540070.1 | 2 - 3 | 1852 | 113 | |
XM_017001768.1 | XP_016857257.1 | 2 - 3 | 1852 | 113 | |
XM_017001769.1 | XP_016857258.1 | 2 - 3 | 2111 | 113 | |
GenBank mRNA | AK002131.1 | - | 1 - 2 | 793 | 113 |
AL834466.1 | - | 1 - 2 | 770 | 113 | |
AY190122.1 | - | 2 - 3 | 1842 | 113 | |
BC008115.1 | - | 1 - 2 | 994 | 113 |