The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040428.3 | NP_001035518.1 | 1 - 2 | 199 | 81 |
NM_018418.4 | NP_060888.2 | 1 - 2 | 199 | 81 | |
XM_005267851.1 | XP_005267908.1 | 1 - 2 | 170 | 81 | |
XM_005267852.1 | XP_005267909.1 | 1 - 2 | 195 | 81 | |
XM_005267854.1 | XP_005267911.1 | 1 - 2 | 170 | 81 | |
XM_005267855.1 | XP_005267912.1 | 1 - 2 | 199 | 81 | |
XM_006720204.1 | XP_006720267.1 | 1 - 2 | 170 | 81 | |
XM_006720205.1 | XP_006720268.1 | 1 - 2 | 170 | 81 | |
XM_017021454.1 | XP_016876943.1 | 1 - 2 | 170 | 81 | |
XM_017021455.1 | XP_016876944.1 | 1 - 2 | 199 | 81 | |
GenBank mRNA | AF144487.3 | - | 1 - 2 | 179 | 81 |
AK055864.1 | - | 1 - 2 | 121 | 81 | |
AK225746.1 | - | 1 - 3 | 170 | 81 | |
AK290631.1 | - | 1 - 2 | 170 | 81 | |
AK293007.1 | - | 1 - 3 | 152 | 81 | |
AL136604.1 | - | 1 - 2 | 179 | 81 | |
BC090875.1 | - | 2 - 3 | 582 | 81 | |
CR936777.1 | - | 1 - 2 | 145 | 81 | |
DA568174.1 | - | 1 - 2 | 199 | 81 | |
EU668353.1 | - | 1 - 2 | 175 | 81 |