Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNTG2
Entrez Gene ID
54221
Gene Name
syntrophin gamma 2
Gene Aliases
G2SYN, SYN5
Chromosome Location
Chr.2: 950868 - 1367991 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018968.3 NP_061841.2 12 - 13 1132 59
XM_011510366.2 XP_011508668.1 10 - 11 1169 59
XM_017004361.1 XP_016859850.1 12 - 13 1094 59
XM_017004362.1 XP_016859851.1 12 - 13 1100 59
XM_017004363.1 XP_016859852.1 12 - 13 1281 59
XM_017004364.1 XP_016859853.1 9 - 10 4422 59
XM_017004365.1 XP_016859854.1 12 - 13 1095 59
XM_017004366.1 XP_016859855.1 12 - 13 1095 59
XM_017004367.1 XP_016859856.1 13 - 14 1170 59
XM_017004373.1 XP_016859862.1 7 - 8 1736 59
GenBank mRNA AJ003029.1 - 12 - 13 1134 59
BC125251.1 - 6 - 7 680 59

View Full Product Details