The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034023.1 | NP_001029195.1 | 4 - 5 | 513 | 96 |
NM_001034024.1 | NP_001029196.1 | 4 - 5 | 473 | 96 | |
NM_019100.4 | NP_061973.1 | 3 - 4 | 694 | 96 | |
XM_005271037.4 | XP_005271094.1 | 3 - 4 | 684 | 96 | |
XM_005271039.1 | XP_005271096.1 | 4 - 5 | 584 | 96 | |
XM_005271040.1 | XP_005271097.1 | 4 - 5 | 461 | 96 | |
XM_006710771.3 | XP_006710834.1 | 2 - 3 | 363 | 96 | |
XM_011541784.2 | XP_011540086.1 | 4 - 5 | 518 | 96 | |
XM_011541785.2 | XP_011540087.1 | 4 - 5 | 538 | 96 | |
XM_017001807.1 | XP_016857296.1 | 4 - 5 | 569 | 96 | |
XM_017001808.1 | XP_016857297.1 | 4 - 5 | 572 | 96 | |
GenBank mRNA | AB037846.1 | - | 4 - 5 | 478 | 96 |
AF265228.1 | - | 3 - 4 | 393 | 96 | |
AK021605.1 | - | 4 - 5 | 457 | 96 | |
AK289366.1 | - | 4 - 5 | 496 | 96 | |
AL137200.1 | - | 4 - 5 | 477 | 96 | |
BC002855.2 | - | 4 - 5 | 462 | 96 | |
BC008053.2 | - | 3 - 4 | 656 | 96 |